Learn More
anti-TRAIL/TNFSF10, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP223644
 View more versions of this product
Catalog No. NBP223644
Add to cart
Description
Description
TRAIL/TNFSF10 Polyclonal antibody specifically detects TRAIL/TNFSF10 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
Specifications
TRAIL/TNFSF10
1 mg/mL
Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry-Paraffin 20 ug/ml
Purified
TNFSF10
Antibody was raised against a peptide corresponding to 17 amino acids near the carboxy terminus of human TRAIL. The immunogen is located within the last 50 amino acids of Trail. Amino Acid Squence: TNEHLIDMDHEASFFGA
35 kDa
0.1 mg
Apoptosis, Cancer, Cell Biology, Signal Transduction, Tumor Suppressors
Primary
8743
Western Blot, ELISA, Immunohistochemistry (Paraffin)
Unconjugated
PBS with 0.02% Sodium Azide
Apo-2 ligand, APO2L, Apo-2Ltumor necrosis factor (ligand) family, member 10, APO2Ltumor necrosis factor apoptosis-inducing ligand splice variant delta, CD253, CD253 antigen, Protein TRAIL, TL2, TNF-related apoptosis-inducing ligand, TRAILTNF-related apoptosis inducing ligand TRAIL, tumor necrosis factor (ligand) superfamily, member 10, tumor necrosis factor ligand superfamily member 10
Rabbit
IgG
Peptide affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human