Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TICAM2 Rabbit anti-Human, Non-human primate, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523115

Catalog No. PIPA523115


Add to cart

Description

Description

TRAM, or TRIF-related adaptor molecule (TICAM2), plays an essential role in the MyD88-independent signaling of TLR4. The functional absence of TRAM in vascular endothelial cells restricts TLR4 signaling to MyD88-dependent pathway. Overexpression of TRAM cDNA in endothelial cells reconstituted LPS-induced Trif-dependent NF-kB activation and cxcl10 promoter activity.
Specifications

Specifications

TICAM2
Polyclonal
Unconjugated
TICAM2
Liquid
TICAM2
Synthetic peptide (87 EDDTDEALRVQNLLQDD 103) of TRAM protein.
Protein A
RUO
Antibody
Polyclonal
Human, Non-human primate
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q86XR7
TIRAP3, TIRP, TRAM, TIR domain-containing adapter molecule 2, Putative NF-kappa-B-activating protein 502, TRIF-related adapter molecule, Toll-like receptor adaptor protein 3, Toll/interleukin-1 receptor domain-containing protein, MyD88-4
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
353376
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit