Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAM2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525887

Catalog No. PIPA525887


Add to cart

Specifications

Specifications

TRAM2
Polyclonal
PBS with 0.09% sodium azide
Q15035
TRAM-like protein, translocating chain-associated membrane protein 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9697
Immunohistochemistry, Western Blot
Unconjugated
TRAM2
Liquid
TRAM2
KLH conjugated synthetic peptide between 342-370 amino acids from the C-terminal region of human TRAM2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit