Learn More
anti-TRANCE/TNFSF11/RANK L, Polyclonal, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP177896
Catalog No. NBP177896
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
TRANCE/TNFSF11/RANK L
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with No Preservative
Delipidation and Defibrination
TNFSF11
This IgG fraction antibody was prepared from rabbit antiserum after repeated immunizations with recombinant truncated mouse RANKL protein (aa 143-316) produced in E.coli.
Ion exchange chromatography
RUO
Store at 4C. Do not freeze.
Polyclonal
Mouse
ELISA, Western Blot
Western Blot 1:1000, ELISA 1:10000
NC_000013.10
CD254, CD254 antigen, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11
Rabbit
IgG
0.1 mg
Cancer
Primary
8600