Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E2F2, Polyclonal, Invitrogen™

Catalog No. PIPA125711
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA125711 50 μg
1 options

Catalog No. PIPA125711

Supplier: Thermo Scientific PA125711

Chicken Polyclonal Antibody

E2F's are DNA binding proteins that associate with negative regulators, such as the retinoblastoma p107 protein, resulting in an altered rate of gene transcription. E2F 2 is regulated by complex formation with Rb p110. E2F family members bind DNA as heterodimers with members of the DP family of polypeptides.

PA1-25711 detects E2F2 from human and mouse samples.PA1-25711 has been successfully used in Western blot procedures.The PA1-25711 immunogen is a synthetic peptide corresponding to the C terminal residues 348-437 of human E2F2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Transcription Factor E2F2
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Gene E2F2
Gene Accession No. Q14209, P56931
Gene Alias E2F-2
Gene Symbols E2F2
Host Species Chicken
Immunogen Synthetic peptide corresponding to the C terminal residues 348-437 of human E2F2.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1870
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.