Learn More
Transferrin Rabbit anti-Human, Biotin, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA127251
Catalog No. PIPA127251
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA1-27255 detects Penicillinase from Enterobacter cloacae samples. PA1-27255 has been successfully used in ELISA, immunomicroscopy and Western blot procedures. The PA1-27255 immunogen is full length protein from E. cloacae.
Transferrins (Tf) are an approximately 80kDa blood plasma glycoprotein synthesized by the liver. They are iron binding transport proteins which can bind two Fe(3+) ions in association with the binding of an anion, usually bicarbonate. It is responsible for the transport of iron from sites of absorption and heme degradation to those of storage and utilization. Transferrin is the primary blood iron transport protein and under normal conditions; approximately one-third of total blood transferrin contains bound iron. Measurement of blood transferrin levels can be used as an indicator for blood iron-carrying capacity and abnormalities of iron metabolism such as anaemia, iron overload and haemochromatosis. Serum transferrin may also have a further role in stimulating cell proliferation.
Specifications
Specifications
Transferrin
Polyclonal
Biotin
TF
Liquid
TF
Transferrin isolated from human serum.
IgG fraction
RUO
Antibody
Polyclonal
Human
ELISA, Immunohistochemistry, Immunomicroscopy, Western Blot
10 mg/mL
0.02M potassium phosphate with 0.15M NaCl, 10mg/mL PEG and 0.01% sodium azide; pH 7.2
P02787
DKFZp781D0156, PRO1557, PRO2086
Rabbit
IgG
1 mg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
7018