Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transferrin Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA3913

Catalog No. 01-674-576


Add to cart

Description

Description

PA3-913 detects transferrin from human and mouse samples. PA3-913 has been successfully used in Western Blot and ICC/IF procedures. The PA3-913 immunogen is human transferrin isolated from plasma.

Transferrins (Tf) are an approximately 80kDa blood plasma glycoprotein synthesized by the liver. They are iron binding transport proteins which can bind two Fe(3+) ions in association with the binding of an anion, usually bicarbonate. It is responsible for the transport of iron from sites of absorption and heme degradation to those of storage and utilization. Transferrin is the primary blood iron transport protein and under normal conditions; approximately one-third of total blood transferrin contains bound iron. Measurement of blood transferrin levels can be used as an indicator for blood iron-carrying capacity and abnormalities of iron metabolism such as anaemia, iron overload and haemochromatosis. Serum transferrin may also have a further role in stimulating cell proliferation.
Specifications

Specifications

Transferrin
Polyclonal
whole serum with 0.05% sodium azide
P02787, Q921I1
DKFZp781D0156, PRO1557, PRO2086
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
TF
Liquid
TF, Trf
Human transferrin isolated from plasma.
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
22041, 7018
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit