Learn More
Transferrin Receptor Mouse anti-Human, Clone: 236-15375, Invitrogen™ Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen A11130
Catalog No. A11130
Add to cart
Description
Description
The anti-human transferrin receptor antibody should be useful for the localization of transferrin receptors by immunofluorescence techniques. This antibody recognizes the human transferrin receptor and does not block binding of transferrin to the receptor. Although fluorescent transferrin conjugates have been widely used to study transferrin receptor-mediated endocytosis, antibodies to the transferrin receptor have proven to be useful for localizing transferrin receptors, studying endocytic recycling pathways and performing enzyme-linked immunosorbent assays (ELISAs). Upon receipt, store desiccated at -20°C. Stored properly, the antibody should remain stable for at least one year. To prepare a stock solution, reconstitute the antibody in PBS with 1% BSA and 2 mM sodium azide. Reconstituted solutions should remain stable for 6 months when stored at 4°C. For longer storage, divide into aliquots and freeze at -20°C.
CD71 (transferrin receptor) is a type II transmembrane glycoprotein expressed as a homodimer in erythroid blood cell line and in activated leukocytes. Upon binding of holotransferrin (complex of transferrin, a monomeric serum glycoprotein (MW~80,000 daltons), and up to two Fe3+ atoms), CD71 is internalized by clathrin-mediated endocytosis. Acidification of endosomes by vesicular membrane proton pumps leads to dissociation of the iron ions, whereas transferrin (apotransferrin) remains associated with CD71 and recycles to the cell surface, where it is released upon exposure to normal pH. CD71 is also involved in uptake of non-transferrin bound iron.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Transferrin Receptor
Monoclonal
Unconjugated
TFRC
Lyophilized
TFRC
human Transferrin Receptor
Protein A
RUO
Antibody
Monoclonal
Human
Immunocytochemistry, Immunofluorescence
236-15375
Antibody with no preservative
P02786
CD71, T9, TFR, TFR1, TR, TRFR, p90, transferrin receptor protein 1
Mouse
IgG1
50 μg
-20°C
Primary
7037