Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transferrin Receptor Mouse anti-Human, Unlabeled, Clone: 2, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612125

 View more versions of this product

Catalog No. BDB612125


Add to cart

Description

Description

Iron acquisition is a prerequisite for cell proliferation. The transferrin receptor is the primary mode of iron uptake. This receptor mediates iron uptake through internalization and recycling of the iron carrying serum protein transferrin. Binding of transferrin to its receptor causes rapid internalization into an acidic intracellular compartment where iron dissociates from transferrin. Both the receptor and transferrin are then recycled through the exocytic pathway. The transferrin receptor is expressed in most cells and tissues, especially proliferating cells, and the receptor is expressed as a dimer of identical intergral membrane proteins. Transferrin receptor mRNA expression is inversely proportional to iron availability, and this regulation occurs through post-transcriptional control of mRNA stability. Transferrin receptor mRNA contains iron responsive elements (IRE) that are bound by IRE-binding proteins (IRP). The RNA binding of IRPs is dependent on iron levels. Thus, iron deprivation leads to increased RNA binding, which masks the recognition site of endonucleases and stabilizes transferrin receptor mRNA.

Western Blotting

Specifications

Specifications

Transferrin Receptor
2
Unconjugated
Affinity Purified
Human Transferrin Receptor aa. 517- 623
150μg
Cell Biology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.