Learn More
TGM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA516272
Catalog No. PIPA516272
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA5-16272 targets Transglutaminase II in IHC (P), IP, and WB applications and shows reactivity with Human, mouse, and Rat samples. The PA5-16272 immunogen is recombit full length human TGase II.
TGase II is implicated in programmed cell death, signal transduction, drug-resistance, cell growth, endocytosis, insulin secretion, cell adhesion, cataract formation, and wound healing.
Specifications
Specifications
TGM2
Polyclonal
Unconjugated
TGM2
Liquid
TGM2
Recombit full length human TGase II
Protein A
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
1 mg/mL
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
P21980, P21981
C polypeptide, TG(C), TGase C, TGase H, TGase-2, TGase-H, protein-glutamine gamma-glutamyltransferase 2, protein-glutamine-gamma-glutamyltransferase, tissue transglutaminase, transglutaminase C, transglutaminase H, transglutaminase-2
Rabbit
IgG
500μL
4° C
Primary
21817, 56083, 7052