Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGM2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514965

Catalog No. PIPA514965

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Transglutaminases are enzymes that catalyze the crosslinking of proteins by epsilon-gamma glutamyl lysine isopeptide bonds. While the primary structure of transglutaminases is not conserved, they all have the same amino acid sequence at their active sites and their activity is calcium-dependent. TGM2 acts as a monomer, is induced by retinoic acid, and appears to be involved in apoptosis. It has been identified as the autoantigen implicated in celiac disease.
Specifications

Specifications

TGM2
Polyclonal
PBS with 0.09% sodium azide
P21980
Tissue transglutaminase; TGase C; TGC; TG(C); Transglutaminase-2; TGase-H
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7052
Western Blot
Unconjugated
TGM2
Liquid
TGM2
KLH conjugated synthetic peptide between 413-442 amino acids from the central region of human Transglutaminase (TGM2)
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit