Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAM1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA521955
Description
Recommended positive controls: H1299. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TRAM1 Polyclonal antibody specifically detects TRAM1 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
TRAM1 | |
Polyclonal | |
Unconjugated | |
Tram1 | |
1810049E02Rik; PNAS8; TRAM; Tram1; TRAMP; Translocating chain-associated membrane protein 1; translocating chain-associating membrane protein; translocating chain-associating membrane protein 1; translocation associated membrane protein 1; translocation-associating membrane protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23471, 72265 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q15629, Q91V04 | |
Tram1 | |
Synthetic peptide corresponding to a region within amino acids 310 and 374 of TRAM1 (Uniprot ID#Q15629) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction