Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMED9 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521422

Catalog No. PIPA521422


Add to cart

Description

Description

PA5-21422 targets transmembrane emp24 protein transport domain containing 9 precursor in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-21422 immunogen is a recombit fragment corresponding to a region within amino acids 1 and 230 of TMED9 (Uniprot IDNo.Q9BVK6). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

TMED9
Polyclonal
Unconjugated
TMED9
Liquid
TMED9
Recombit fragment corresponding to a region within amino acids 1 and 230 of TMED9 (Uniprot IDNo.Q9BVK6)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q9BVK6
GMP25, HSGP25L2G, p25, glycoprotein 25L2, gp25L2 protein, p24 family protein alpha-2, p24alpha2, transmembrane emp24 domain-containing protein 9
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
54732
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit