Learn More
Transmissible Gastroenteritis Virus Mouse anti-Virus, Clone: TGEV15-948.2.1, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA110786
Catalog No. 01-671-816
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-10786 detects Transmissible Gastroenteritis Virus. MA1-10786 has been successfully used in ELISA and neutralization applications. The MA1-10786 immunogen is TGEV-infected ST cells.
Specifications
Specifications
Transmissible Gastroenteritis Virus
Monoclonal
1.28 mg/mL
PBS with no preservative; pH 7.4
TGV
TGEV-infected ST cells.
Purified
RUO
Antibody
Monoclonal
ELISA
TGEV15-948.2.1
Unconjugated
Liquid
Mouse
IgG2a
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Virus