Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tartrate Resistant Acid Phosphatase, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA521317
Description
PA5-21317 targets TRAP in WB and IHC-P applications and shows reactivity with Human and mouse samples.The PA5-21317 immunogen is a recombinant fragment corresponding to a region within amino acids 28 and 325 of TRAP (Uniprot ID#P13686).Specifications
Tartrate Resistant Acid Phosphatase | |
Polyclonal | |
Unconjugated | |
P13686, Q05117 | |
ACP5 | |
Recombinant fragment corresponding to a region within amino acids 28 and 325 of TRAP (Uniprot ID#P13686) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
1mg/mL | |
ACP5 | |
SPENCDI, TRAP, TrATPase, tartrate-resistant acid ATPase, tartrate-resistant acid phosphatase type 5, TRAPase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11433, 54 | |
Store at -20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction