Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAPPC3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523684

Catalog No. PIPA523684

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, chicken, mouse, porcine, non-human primate and rat based on sequence homology.

TRAPPC3 is a component of the TRAPP complex, which isinvolved in tethering of transport vesicles to the cis-Golgimembrane.
Specifications

Specifications

TRAPPC3
Polyclonal
PBS with 0.09% sodium azide
O43617
BET3, 1110058K12Rik, BET3 homolog, trafficking protein particle complex subunit 3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
27095
Flow Cytometry, Western Blot
Unconjugated
TRAPPC3
Liquid
TRAPPC3
KLH conjugated synthetic peptide between 17-46 amino acids from the N-terminal region of human TRAPPC3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit