Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAPPC4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA511522

Catalog No. PIPA511522

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

TRAPPC4 is part of the multisubunit TRAPP (transport protein particle) complex and interacts with SDC2. It may play a role in vesicular transport from endoplasmic reticulum to Golgi. TRAPP proteins are involved in tethering during vesicle transport.
Specifications

Specifications

TRAPPC4
Polyclonal
PBS with 0.09% sodium azide
Q9Y296
Synbindin; TRS23 homolog; Hematopoietic stem/progenitor cell protein 172; SBDN
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51399
Immunohistochemistry, Western Blot
Unconjugated
TRAPPC4
Liquid
TRAPPC4
KLH conjugated synthetic peptide between 54-84 amino acids from the N-terminal region of human TRAPPC4
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit