Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAPPC9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NB10055735
Description
TRAPPC9 Polyclonal specifically detects TRAPPC9 in Human, Mouse, Bovine, Canine, Chicken, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
TRAPPC9 | |
Polyclonal | |
Western Blot 1:100-1:2000. Use reported in scientific literature (Zahoor et al (2010)), Immunocytochemistry/Immunofluorescence reported in scientific literature (Zahoor et al (2010)), Immunoprecipitation 1:10-1:500. Use reported in scientific literature (Zahoor et al (2010)) | |
Q96Q05 | |
TRAPPC9 | |
Amino acids 282-299 (KKDFVGLDTDSRHYKKRC) of human NIBP was used as the immunogen. | |
0.1 mL | |
Primary | |
The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_A and CRA_b), chimpanzee, monkey, mouse, opossum, rat and dog and 94% homologous in xenopus. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Whole Rabbit antisera with 50% Glycerol with 0.025% Sodium Azide | |
IKBKBBP, IKK2 binding protein, KIAA1882IBP, MRT13MGC4769, NIBPMGC4737, NIK- and IKBKB-binding protein, NIK and IKK-beta binding protein, T1, trafficking protein particle complex 9, trafficking protein particle complex subunit 9, TRAPP 120 kDa subunit, TRS120, Tularik gene 1 protein | |
Rabbit | |
Unpurified | |
RUO | |
83696 | |
Human, Mouse, Bovine, Canine, Chicken, Primate | |
Antisera |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction