Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Anti- Trastuzumab (Anti-Idiotype) Alexa Fluor™ 750-conjugated Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB95471S100UG
This item is not returnable.
View return policy
Description
Trastuzumab Monoclonal specifically detects Trastuzumab in Multi-Species samples. It is validated for Flow Cytometry.Specifications
Trastuzumab | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide | |
Full-length Trastuzumab | |
100 μg | |
Detects Trastuzumab in Western blots and direct ELISAs. In direct ELISAs, no cross-reactivity with Cetuximab, Ibritumomab, Rituximab, Adalimumab, Tositumomab, Brentuximab, Gemtuzumab, Solanezumab, Teprotumumab, and Aducanumab is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG |
Flow Cytometry | |
2261E | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Multi-species | |
Supernatant |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction