Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRBP Mouse anti-Human, Mouse, Clone: 46D1, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific LFMA0209
Description
A suggested positive control for this product is 293T cells.
HIV-1, the causative agent of acquired immunodeficiency syndrome (AIDS), contains an RNA genome that produces a chromosomally integrated DNA during the replicative cycle. Activation of HIV-1 gene expression by the transactivator Tat is dependent on an RNA regulatory element (TAR) located downstream of the transcription initiation site. The protein encoded by this gene binds between the bulge and the loop of the HIV-1 TAR RNA regulatory element and activates HIV-1 gene expression in synergy with the viral Tat protein. Alternative splicing results in multiple transcript variants encoding different isoforms. This gene also has a pseudogene.Specifications
TRBP | |
Monoclonal | |
1 mg/mL | |
TARBP2 | |
LOQS, TRBP, TRBP1, TRBP2 | |
Mouse | |
Protein G | |
RUO | |
21357, 6895 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot | |
46D1 | |
Unconjugated | |
Q15633, P97473 | |
TARBP2 | |
Recombit human protein purified from E.coli(GST-TRBP) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title