Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRBP Rabbit anti-Equine, Human, Non-human primate, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523175

Catalog No. PIPA523175


Add to cart

Description

Description

TRBP is an RNA associated nuclear protein.  It contains 3 DRBM (double stranded RNA-binding) domains and plays an important role by binding to HIV-1 TAR RNA located in the long terminal repeat (LTR). TRBP acts synergistic with viral TAT protein in HIV-1 expression activation. It binds preferentially to TAR and other double-stranded RNAs with G+C-rich characteristics. It is important in regulation of transcription from RNA polymerase II promoter and is widely expressed in most of the tissues.
Specifications

Specifications

TRBP
Polyclonal
Unconjugated
TARBP2
Liquid
TARBP2
Synthetic peptide (344 REAARGEAARRALQ 357) of TRBP protein.
Protein G
RUO
Antibody
Polyclonal
Equine, Human, Non-human Primate
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.2% gelatin and 0.05% sodium azide
Q15633
LOQS, TRBP, TRBP1, TRBP2, RISC-loading complex subunit TARBP2, TAR (HIV) RNA binding protein 2, TAR (HIV) RNA-binding protein 2, TAR (HIV) RNA-binding protein TRBP1, TAR RNA binding protein 2, TAR RNA-binding protein 2, trans-activation responsive RNA-binding protein, trans-activation-responsive RNA-binding protein
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100062193, 6895
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit