Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TREF1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526365

Catalog No. PIPA526365


Add to cart

Description

Description

TREF1 encodes a zinc-finger transcriptional regulating protein which interacts with CBP/p300 to regulate the human gene CYP11A1.
Specifications

Specifications

TREF1
Polyclonal
PBS with 0.09% sodium azide
Q96PN7
BCAR2, HSA277276, RAPA, RP1-139D8.5, TReP-132, dJ139D8.5, breast cancer anti-estrogen resistance 2, rapa-1, rapa-2, transcriptional regulating protein 132, transcriptional-regulating factor 1, transcriptional-regulating protein 132, zinc finger protein rapa, zinc finger transcription factor TReP-132
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55809
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
TRERF1
Liquid
TRERF1
KLH conjugated synthetic peptide between 1136-1164 amino acids from the C-terminal region of human TREF1
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit