Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TREM2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525383

Catalog No. PIPA525383


Add to cart

Description

Description

TREM2 is a membrane protein that forms a receptor signaling complex with TYROBP. The encoded protein may be involved in chronic inflammation by triggering the production of constitutive inflammatory cytokines. Defects in this gene are a cause of polycystic lipomembranous osteodysplasia with sclerosing leukoencephalopathy (PLOSL).
Specifications

Specifications

TREM2
Polyclonal
PBS with 0.09% sodium azide
Q9NZC2
TREM-2, Trem2a, Trem2b, Trem2c, triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2a
Rabbit
IgG
200μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54209
Western Blot
Unconjugated
TREM2
Liquid
TREM2
KLH conjugated synthetic peptide between 22-50 amino acids from the N-terminal region of human TREM2
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit