Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

TREM3 Rat anti-Mouse, Clone: 295502, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA524003

Catalog No. PIMA524003



Description

Description

In direct ELISAs and Western blots, no cross-reactivity with recombit mouse (rm) TREM-1 or rmTREM-2b is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

Specifications

Specifications

TREM3
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Lyophilized
Rat
IgG2A
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
58218
Western Blot
295502
Unconjugated
Trem3
Trem3
Mouse myeloma cell line NS0-derived recombit mouse TREM-3 Met1-Pro135 (predicted)
Protein A/G
RUO
Antibody
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit