three prime repair exonuclease 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against a full-length human TREX1 protein.

Manufacturer: Abnova Corporation H00011277D01P

Catalog No. 89-018-079

  Check Availability
Add to Cart

    Specifications

    Formulation In 1X PBS, pH 7.4
    Gene TREX1
    Gene Accession No. NM_016381
    Gene Alias AGS1, AGS5, CRV, DKFZp434J0310, DRN3, HERNS
    Gene Symbols TREX1
    Applications Western Blotting
    Species Reactivity Human, Mouse
    Storage Requirements Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
    Gene ID (Entrez) 11277
    Description three prime repair exonuclease 1
    Host Species Rabbit
    Quantity 100μg
    Regulatory Status RUO
    Antigen three prime repair exonuclease 1
    Format Affinity Purified
    Immunogen TREX1 (NP_057465.1, 1 a.a. ∽369 a.a) full-length human protein.
    Purification Method Protein A column
    Primary or Secondary Primary
    Monoclonal or Polyclonal Polyclonal
    Conjugate Unlabeled
    Cross Reactivity Human,Mouse

    This gene encodes the major 3'->5' DNA exonuclease in human cells. The protein is a nonprocessive exonuclease that may serve a proofreading function for a human DNA polymerase. It is also a component of the SET complex, and acts to rapidly degrade 3' ends of nicked DNA during granzyme A-mediated cell death. Mutations in this gene result in Aicardi-Goutieres syndrome, chilblain lupus, and Cree encephalitis. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

    Sequence: MGPGARRQGRIVQGRPEMCFCPPPTPLPPLRILTLGTHTPTPCSSPGSAAGTYPTMGSQALPPGPMQTLIFFDMEATGLPFSQPKVTELCLLAVHRCALESPPTSQGPPPTVPPPPRVVDKLSLCVAPGKACSPAASEITGLSTAVLAAHGRQCFDDNLANLLLAFLRRQPQPWCLVAHNGDRYDFPLLQAELAMLGLTSALDGAFCVDSITALKALERASSPSEHGPRKSYSLGSIYTRLYGQSPPDSHTAEGDVLALLSICQWRPQALLRWVDAHARPFGTIRPMYGVTASARTKPRPSAVTTTAHLATTRNTSPSLGESRGTKDLPPVKDPGALSREGLLAPLGLLAILTLAVATLYGLSLATPGE

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit