Learn More
three prime repair exonuclease 1, Mouse, Polyclonal Antibody, Abnova
Mouse polyclonal antibody raised against a full-length human TREX1 protein.
Manufacturer: Abnova Corporation H00011277B02
Description
This gene encodes the major 3'->5' DNA exonuclease in human cells. The protein is a nonprocessive exonuclease that may serve a proofreading function for a human DNA polymerase. It is also a component of the SET complex, and acts to rapidly degrade 3' ends of nicked DNA during granzyme A-mediated cell death. Mutations in this gene result in Aicardi-Goutieres syndrome, chilblain lupus, and Cree encephalitis. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: MGPGARRQGRIVQGRPEMCFCPPPTPLPPLRILTLGTHTPTPCSSPGSAAGTYPTMGSQALPPGPMQTLIFFDMEATGLPFSQPKVTELCLLAVHRCALESPPTSQGPPPTVPPPPRVVDKLSLCVAPGKACSPAASEITGLSTAVLAAHGRQCFDDNLANLLLAFLRRQPQPWCLVAHNGDRYDFPLLQAELAMLGLTSALDGAFCVDSITALKALERASSPSEHGPRKSYSLGSIYTRLYGQSPPDSHTAEGDVLALLSICQWRPQALLRWVDAHARPFGTIRPMYGVTASARTKPRPSAVTTTAHLATTRNTSPSLGESRGTKDLPPVKDPGALSREGLLAPLGLLAILTLAVATLYGLSLATPGESpecifications
three prime repair exonuclease 1 | |
Unconjugated | |
No additive | |
NM_016381 | |
AGS1|AGS5|CRV|DKFZp434J0310|DRN3|HERNS | |
Mouse | |
50 uL | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Primary | |
11277 |
Western Blot | |
Mouse polyclonal antibody raised against a full-length human TREX1 protein. | |
TREX1 | |
Serum | |
TREX1 | |
TREX1 (NP_057465, 1 a.a. ~ 369 a.a) full-length human protein. | |
RUO | |
Y | |
Polyclonal | |
Human |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.