Learn More
anti-TREX1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP229617
 View more versions of this product
Catalog No. NBP229617
Add to cart
Description
Description
TREX1 Polyclonal specifically detects TREX1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
TREX1
Polyclonal
Unconjugated
PBS with 0.05% Sodium Azide
3'-5' exonuclease TREX1, AGS1, Aicardi-Goutieres syndrome 1, CRV, DKFZp434J0310, DNase III, DRN3, EC 3.1.11.2, HERNS, three prime repair exonuclease 1,3' repair exonuclease 1
Rabbit
IgG purified
RUO
Primary
Human
Purified
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
1.0 mg/mL
Western Blot 1-2 ug/ml, Immunohistochemistry 5-10 ug/ml, Immunohistochemistry-Paraffin 5-10 ug/ml, Immunohistochemistry-Frozen 5-10 ug/ml
Q9NSU2
TREX1
Partial recombinant protein made to an internal portion of human TREX1 protein (between residues 200-350) [NP_057465.1]
0.1 mg
DNA Repair, Editing and Processing Endonucleases
11277
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG