Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRHDE Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525545

Catalog No. PIPA525545


Add to cart

Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

This gene encodes a member of the peptidase M1 family. TRHDE is an extracellular peptidase that specifically cleaves and inactivates the neuropeptide thyrotropin-releasing hormone.
Specifications

Specifications

TRHDE
Polyclonal
Unconjugated
TRHDE
Liquid
TRHDE
KLH conjugated synthetic peptide between 951-980 amino acids from the C-terminal region of human TRHDE
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q9UKU6
PAP-II, PGPEP2, TRH-DE, TRH-degrading ectoenzyme, TRH-specific aminopeptidase, pyroglutamyl-peptidase II, thyroliberinase, thyrotropin-releasing hormone degrading ectoenzyme, thyrotropin-releasing hormone-degrading ectoenzyme
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
29953
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit