Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H3K4me3 Monoclonal Antibody (G.532.8), Invitrogen™
Rabbit Monoclonal Antibody
Supplier: Thermo Scientific MA511199
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with methylated histone H3 Lys9, Lys27, Lys36 or methylated histone H4 Lys20.
H3K4me3 Monoclonal antibody specifically detects H3K4me3 in Drosophila, Human, Mouse, Non-human Primate, Rat, Yeast samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Peptide Array, Western Blot.Specifications
H3K4me3 | |
Monoclonal | |
121 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
P61830, P68431, P68433, P84228, Q71DI3 | |
H3C1, H3C10, H3C12, H3C14, H3C15, H3C2, H3C7, HHT1, His3:CG31613 | |
Synthetic peptide corresponding to the amino terminus of histone H3 in which Lys4 is tri-methylated | |
100 μL | |
Primary | |
Drosophila, Human, Mouse, Non-human Primate, Rat, Yeast | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Peptide Array, Western Blot | |
G.532.8 | |
Unconjugated | |
H3C12 | |
B0035.7; BUR5; CELE_T10C6.12; CG31613; CG31613-PA; CG33803; CG33806; CG33809; CG33812; CG33815; CG33818; CG33821; CG33824; CG33827; CG33830; CG33833; CG33836; CG33839; CG33842; CG33845; CG33848; CG33851; CG33854; CG33857; CG33860; CG33863; CG33866; Dmel\CG31613; Dmel_CG31613; F07B7.10; F07B7.3; F08G2.2; F17E9.13; F35H10.1; F45F2.4; F54E12.5; F55G1.10; H02I12.7; H3; H3 histone; H3 histone family, member A; H3 histone family, member I; H3 histone family, member K; H3 histone family, member L; H3 histone family, member M; H3 histone, family 2; H3 histone, family 3A; H3 histone, family 3B; H3 histone, family 3B (H3.3B); H3 histone, family 3B.1; H3 histone, family 3C; H3 K4; H3.1-221; H3.1-291; H3.1-I; H3.2; H3.2-221; H3.2-614; H3.2-615; H3.2-616; h3.2a; H3.3A; H3.3B; H3.5; H3/A; H3/b; H3/d; H3/f; H3/i; H3/j; H3/k; H3/l; H3/M; H3/n; H3/o; H3-143; H3-291; H3-3A; H3-3b; h3-5; H3-53; H3-614; H3a; H3b; H3-B; H3C1; H3C10; H3C11; H3C12; H3C2; H3C3; H3C4; H3C6; H3C7; H3c8; H3f; H3-F; H3F1K; H3F2; | |
Rabbit | |
Affinity chromatography | |
RUO | |
126961, 260423, 318847, 319150, 319152, 333932, 360198, 8350, 8356, 8357, 8358, 852295, 8968, 97114 | |
-20°C | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction