Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tri-Methyl-Histone H3 (Lys4) Rabbit anti-Human, Many, Mouse, Plant, Zebrafish, Polyclonal, Invitrogen
Rabbit Polyclonal Antibody
Manufacturer: Invitrogen 491005
Description
Histone H3 is one of the DNA-binding proteins found in the chromatin of all eukaryotic cells. H3 along with four core histone proteins binds to DNA forming the structure of the nucleosome. Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Post tranlationally, histones are modified in a variety of ways to either directly change the chromatin structure or allow for the binding of specific transcription factors. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of post-translational modification that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.Specifications
Tri-Methyl-Histone H3 (Lys4) | |
Polyclonal | |
Unconjugated | |
HIST1H3A | |
Liquid | |
H3c1, HIST1H3A | |
Raised against the region of the histone H3 containing the trimethylated lysine 4 (or [K4me3]), using a KLH-conjugated synthetic peptide. | |
Affinity chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Many, Mouse, Plant, Zebrafish |
ELISA, Immunocytochemistry, Immunofluorescence, Peptide array, Peptide Array, Western Blot | |
1.4 mg/mL | |
PBS with 0.05% Proclin 300, 0.05% sodium azide | |
P68431, P68433 | |
H3K4me3, Histone H3, Histone 1, H3a, Histone H3.1, histone H3/a, HIST1H3A | |
Rabbit | |
IgG | |
50 μg | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
360198, 8350 |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title