Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIB3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA515480

Catalog No. PIPA515480


Add to cart

Description

Description

NPK is a putative protein kinase that is induced by the transcription factor NF-kappaB. The encoded protein is a negative regulator of NF-kappaB and can also sensitize cells to TNF- and TRAIL-induced apoptosis. In addition, this protein can negatively regulate the cell survival serine-threonine kinase AKT1.
Specifications

Specifications

TRIB3
Polyclonal
Unconjugated
TRIB3
Liquid
TRIB3
KLH conjugated synthetic peptide between 301-331 amino acids from the C-terminal region of human TRIB3
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
2 mg/mL
PBS with 0.09% sodium azide
Q96RU7, Q8K4K2
Neuronal cell death inducible putative kinase; SKIP3; TRIB3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
228775, 57761
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit