Learn More
Trichomonas foetus Mouse anti-Protozoa, Clone: H63J, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA518269
Catalog No. PIMA518269
Add to cart
Description
Description
This antibody does not cross-react with T. vaginalis. Purity is >90%. Recommended pairs for ELISA: MA5-18269 (capture) and MA5-18270 (conjugated, detection).
Specifications
Specifications
Trichomonas foetus
Monoclonal
100 μg/mL
PBS with 0.1% sodium azide; pH 7.2
T. foetus
Trichomonas foetus native protein
Protein A
RUO
Antibody
Monoclonal
ELISA, Immunofluorescence
H63J
Unconjugated
Liquid
Mouse
IgG2b
100 μg
4° C
Primary
Protozoa