Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIF Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA120824

Catalog No. PA120824


Add to cart

Description

Description

PA1-20824 detects TRIF in human and mouse samples. PA1-20824 has been successfully used in Western Blot procedures. By Western blot, PA1-20824 detects the ~ 66 kDa TRIF protein. PA1-20824 immunogen is a Synthetic peptide, corresponding to amino acids 143-158 of Mouse TRIF.

TRIF plays an essential role in the MyD88-independent signaling of TLR3.
Specifications

Specifications

TRIF
Polyclonal
whole serum diluted in PBS with 0.2% gelatin and 0.02% sodium azide
Q8IUC6, Q80UF7
hCG_39321, HSD34, RNF36, Trif
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
Unconjugated
TICAM1
Liquid
TICAM1
Synthetic peptide, corresponding to amino acids 143-158 of Mouse TRIF.
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
106759, 148022
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit