Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIL Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523454

Catalog No. PIPA523454


Add to cart

Description

Description

TRIL (TLR4 interactor with leucine rich repeats) is a single-pass type 1 membrane protein and a novel component of the TLR4 complex. TRIL expression and interaction with TLR4 is enhanced by LPS stimulation. Knockdown of TRIL decreases the effect of LPS in cell lines, human PBMC, and glial cells. Expression of TRIL is seen in a number of tissues, most prominently in the brain but also in the spinal cord, lung, kidney, and ovary.
Specifications

Specifications

TRIL
Polyclonal
Unconjugated
TRIL
Liquid
TRIL
Synthetic peptide (797 SLRREDRLLQRFAD 811) of TRIL protein.
Protein A
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q7L0X0
TLR4 interactor with leucine rich repeats
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
9865
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit