Learn More
tripartite motif-containing 10, Mouse, Polyclonal Antibody, Abnova
Mouse polyclonal antibody raised against a full-length human TRIM10 protein.
Manufacturer: Abnova Corporation H00010107B01
Description
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to cytoplasmic bodies. Studies in mice suggest that this protein plays a role in terminal differentiation of erythroid cells. Alternate splicing of this gene generates two transcript variants encoding different isoforms. [provided by RefSeq
Sequence: MASAASVTSLADEVNCPICQGTLREPVTIDCGHNFCRACLTRYCEIPGPDLEESPTCPLCKEPFRPGSFRPNWQLANVVENIERLQLVSTLGLGEEDVCQEHGEKIYFFCEDDEMQLCVVCREAGEHATHTMRFLEDAAAPYREQIHKCLKCLRKEREEIQEIQSRENKRMQVLLTQVSTKRQQVISEFAHLRKFLEEQQSILLAQLESQDGDILRQRDEFDLLVAGEICRFSALIEELEEKNERPARELLTDIRSTLIRCETRKCRKPVAVSPELGQRIRDFPQQALPLQREMKMFLEKLCFELDYEPAHISLDPQTSHPKLLLSEDHQRAQFSYKWQNSPDNPQRFDRATCVLAHTGITGGRHTWVWMARVPGDSGCCQFCSPPSVLGTEVAASpecifications
tripartite motif-containing 10 | |
Unconjugated | |
No additive | |
BC093926.1 | |
HERF1|MGC141979|RFB30|RNF9 | |
Mouse | |
50 uL | |
Ubiquitin/Proteasome | |
Y | |
Polyclonal | |
Human |
Western Blot | |
Mouse polyclonal antibody raised against a full-length human TRIM10 protein. | |
TRIM10 | |
Serum | |
TRIM10 | |
TRIM10 (AAH93926.1, 1 a.a. ~ 395 a.a) full-length human protein. | |
RUO | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Primary | |
10107 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.