Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TRIM21/SSA1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP140338

Catalog No. NBP140338


Add to cart

Description

Description

TRIM21/SSA1 Polyclonal antibody specifically detects TRIM21/SSA1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

TRIM21/SSA1
0.2 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation 5-15 ug/mg lysate
P19474
52 kDa ribonucleoprotein autoantigen Ro/SS-A, 52 kDa Ro/SSA, E3 ubiquitin-protein ligase TRIM21, EC 6.3.2.-, rhTRIM21, RNF81RING finger protein 81, ro(SS-A), RO52SS-A, Sicca syndrome antigen A, Sjoegren syndrome type A antigen, Sjogren syndrome antigen A1 (52kDa, ribonucleoprotein autoantigen SS-A/Ro), SSA1SSA, tripartite motif containing 21,52 kDa Ro protein, tripartite motif-containing 21, Tripartite motif-containing protein 21
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human. Based on 100% sequence identity, this antibody is predicted to react with Rhesus Monkey, Gorilla, Chimpanzee, and Northern white-cheeked gibbon.
Immunoprecipitation, Western Blot
Unconjugated
TBS and 0.1% BSA with 0.09% Sodium Azide
Affinity Purified
TRIM21
The immunogen for this product maps to a region between residue 425 and 475 of human Sjogren syndrome antigen A1 using the numbering given in entry NP_003132.2 (GeneID 6737).
54 kDa
0.1 ml
Zinc Finger
Primary
6737
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only