Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TRIM31, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP213483

Catalog No. NBP213483

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

TRIM31 Polyclonal antibody specifically detects TRIM31 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

TRIM31
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
C6orf13E3 ubiquitin-protein ligase TRIM31, EC 6.3.2.-, HCG1, HCGItripartite motif-containing 31 gamma, RNFring finger protein, tripartite motif containing 31, tripartite motif-containing 31, Tripartite motif-containing protein 31
Rabbit
IgG
0.1 ml
Zinc Finger
Primary
11074
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Affinity Purified
TRIM31
This antibody was developed against Recombinant Protein corresponding to amino acids: DSLKTKQNMPPRQLLEDIKVVLCRSEEFQFLNPTPVPLELEKKLSEAKSR HDSITGSLKKFKDQLQADRKKDENRFFKSMNKNDMKSWGLLQK
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only