Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM36 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528401

Catalog No. PIPA528401


Add to cart

Description

Description

PA5-28401 targets TRIM36 in IF and WB applications and shows reactivity with Human samples. The PA5-28401 immunogen is synthetic peptide corresponding to a region within amino acids 666 and 728 of Human TRIM36. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. Multiple alternatively spliced transcript variants that encode different protein isoforms have been described for this gene.
Specifications

Specifications

TRIM36
Polyclonal
Unconjugated
TRIM36
Liquid
TRIM36
Synthetic peptide corresponding to a region within amino acids 666 and 728 of Human TRIM36
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
PBS with 10% glycerol and 0.01% thimerosal; pH 7
Q9NQ86
HAPRIN, RBCC728, RNF98, E3 ubiquitin-protein ligase TRIM36, RING finger protein 98, tripartite motif protein 36, tripartite motif-containing 36, tripartite motif-containing protein 36, zinc-binding protein Rbcc728
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
55521
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit