Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM37 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521737

Catalog No. PIPA521737


Add to cart

Description

Description

PA5-21737 targets TRIM37 in ICC, IF, IHC (P), and WB applications and shows reactivity with Human samples. The PA5-21737 immunogen is a synthetic peptide corresponding to a region within amino acids 902 and 964 of TRIM37 (Uniprot IDNo.O94972). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

TRIM37
Polyclonal
Unconjugated
TRIM37
Liquid
TRIM37
Synthetic peptide corresponding to a region within amino acids 902 and 964 of TRIM37 (Uniprot IDNo.O94972)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1.02 mg/mL
PBS with 10% glycerol and 0.01% thimerosal; pH 7
O94972
MUL, POB1, TEF3, E3 ubiquitin-protein ligase TRIM37, RING-B-box-coiled-coil protein, mulibrey ism protein, tripartite motif-containing 37, tripartite motif-containing protein 37
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
4591
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit