Learn More
tripartite motif-containing 39 (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human TRIM39 protein.
Supplier: Abnova Corporation H00056658B01
Description
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The function of this protein has not been identified. This gene lies within the major histocompatibility complex class I region on chromosome 6. Alternate splicing results in two transcript variants encoding different isoforms. [provided by RefSeq
Sequence: MAETSLLEAGASAASTAAALENLQVEASCSVCLEYLKEPVIIECGHNFCKACITRWWEDLERDFPCPVCRKTSRYRSLRPNRQLGSMVEIAKQLQAVKRKIRDESLCPQHHEALSLFCYEDQEAVCLICAISHTHRAHTVVPLDDATQEYKEKLQKCLEPLEQKLQEITRCKSSEEKKPGELKRLVESRRQQILREFEELHRRLDEEQQVLLSRLEEEEQDILQRLRENAAHLGDKRRDLAHLAAEVEGKCLQSGFEMLKDVKSTLEKCEKVKTMEVTSVSIELEKNFSNFPRQYFALRKILKQLIADVTLDPETAHPNLVLSEDRKSVKFVETRLRDLPDTPRRFTFYPCVLATEGFTSGRHYWEVEVGDKTHWAVGVCRDSVSRKGELTPLPETGYWRVRLWNGDKYAATTTPFTPLHIKVKPKRVGIFLDYEAGTLSFYNVTDRSHIYTFTDTFTEKLWPLFYPGIRAGRKNAAPLTIRPPTDWESpecifications
tripartite motif-containing 39 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human TRIM39 protein. | |
TRIM39 | |
MGC32984/RNF23/TFP/TRIM39B | |
Mouse | |
50 μL | |
Ubiquitin/Proteasome | |
Primary | |
Human | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
NM_172016.1 | |
TRIM39 | |
TRIM39 (NP_742013.1, 1 a.a. ∼ 488 a.a) full-length human protein. | |
RUO | |
Yes | |
56658 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.