Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TRIM5, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP152454

Catalog No. NBP152454


Add to cart

Description

Description

TRIM5 Polyclonal antibody specifically detects TRIM5 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications

Specifications

TRIM5
1 mg/mL
Western Blot 1-2 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 5 ug/ml
Affinity Purified
TRIM5
Synthetic peptide from monkey TRIM5.
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
synthetic peptide corresponding to amino acids near the mid-region of rhesus monkey TRIM5a
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PBS with 0.02% Sodium Azide
EC 6.3.2, EC 6.3.2.-, RING finger protein 88, RNF88tripartite motif protein TRIM, TRIM5alpha, tripartite motif containing 5, tripartite motif protein TRIM5, tripartite motif-containing 5, tripartite motif-containing protein 5
Rabbit
IgG
0.05 mg
Immunology, Lipid and Metabolism, Virology Bacteria and Parasites, Zinc Finger
Primary
85363
Human, Primate
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only