Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trimethyl Histone H3 (Lys27) Mouse anti-Human, Mouse, AlexaFluor™488, Clone: 18E9.1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051951AF488
Description
This Anti-trimethyl Histone H3 (Lys27) Antibody, clone 18E9.1, Alexa Fluor™ 488 Conjugate is validated for use in immunocytochemistry for the detection of trimethyl Histone H3 (Lys27).Specifications
trimethyl Histone H3 (Lys27) | |
Monoclonal | |
Alexa Fluor 488 | |
NP_002098 | |
Mouse | |
Protein G purified | |
Primary | |
2-8°C for one year |
Immunocytochemistry | |
18E9.1 | |
Purified mouse conjugate in buffer containing PBS, 15mg/mL BSA, and 0.1% Sodium Azide. | |
H3F3B, H3.3B | |
Linear peptide corresponding to trimethyl Histone H3 (Lys27). | |
100 μL | |
Human, Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only