Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trimethyl Histone H3 (Lys27) Mouse anti-Human, Mouse, Clone: 18E9.1, MilliporeSigma™
Mouse Monoclonal Antibody
$145.60 - $610.00
Specifications
Antigen | trimethyl Histone H3 (Lys27) |
---|---|
Clone | 18E9.1 |
Applications | Immunocytochemistry, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
05-195-1SMI
|
MilliporeSigma
051951S |
10 μL |
Each for $145.60
|
|
|||||
05-195-1MI
|
MilliporeSigma
051951 |
100 μL |
Each for $610.00
|
|
|||||
Description
Anti-trimethyl Histone H3 (Lys27) Antibody, clone 18E9.1 is a mouse monoclonal antibody for detection of trimethyl Histone H3 (Lys27) also known as H3K27me3, Histone H3 (tri methyl K27) and has been validated in western blot, immunocytochemistry.Specifications
trimethyl Histone H3 (Lys27) | |
Immunocytochemistry, Western Blot | |
Unconjugated | |
Mouse | |
Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
HIST3H3, H3FT, H3/t, H3t, H3/g, H3.4, H3T | |
IgG2b κ | |
Protein G Purified |
18E9.1 | |
Monoclonal | |
Purified | |
Human, Mouse | |
NP_002098 | |
Linear peptide corresponding to human Histone H3 trimethylated at Lys27. | |
Primary | |
2-8°C for one year |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title