Learn More
Trimethyl-Histone H3 (Lys36) Rabbit anti-Chicken, Clone: MC86, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma 05801
Catalog No. 05-801-MI
Add to cart
Description
Description
Use Anti-trimethyl-Histone H3 (Lys36) Antibody, clone MC86 (rabbit monoclonal antibody) validated in DB, western blot, PIA to detect trimethyl-Histone H3 (Lys36) also known as H3K36me3, Histone H3 (tri methyl K36).
Specifications
Specifications
trimethyl-Histone H3 (Lys36)
MC86
Protein A Purified immunoglobulin in a solution containing 0.07M Tris-glycine, 0.105M NaCl, pH 7.4, 0.035% sodium azide, 30% glycerol.
Purified
peptide containing the sequence GVme3KKP, in which me3K corresponds to trimethyl lysine 36 of human histone H3
Protein A purfied
−20°C for 24 months. Aliquot to avoid freeze/thaw cycles.
Monoclonal
Chicken
Dot Blot, Inhibition Assays, Western Blot
Unconjugated
NP_003484
Rabbit
IgG
100μL
Primary
HIST3H3, H3FT, MGC126886, H3t, MGC126888, H3T, H3/g, H3.4, H3/t