Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-trimethyl-Histone H3 (Lys4), Clone: 15-10C-E4, (Upstate) Non-distribution product as customer accommodation.

Manufacturer:  MilliporeSigma™ 05745R/DEL

Catalog No. 50-171-803

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the 'beads on a string' structure.

The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Host Species: Rabbit
Clone: 15-10C-E4
Isotype: IgG
Species Reactivity: Human
Immunogen: BSA-conjugated, synthetic peptide containing the sequence ...RTme3KQT... in which me3K corresponds to trimethyl-lysine 4 of human histone H3.
Molecular Weight: 17kDa

Applications:

ELISA, Western Blotting

Specifications

Specifications

trimethyl-Histone H3 (Lys4)
Unconjugated
Rabbit
IgG
RUO
Primary
Human
15-10C-E4
Semi-purified
BSA-conjugated, synthetic peptide containing the sequence …RTme3KQT… in which me3K corresponds to trimethyl-lysine 4 of human histone H3
100μL
Epigenetics
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit