Learn More
anti-trimethyl Histone H3 (Lys79), Polyclonal, EMD Millipore™
Manufacturer:  Millipore Sigma 07952
 View more versions of this product
Catalog No. 07-952-MI
Add to cart
Specifications
Specifications
trimethyl Histone H3 (Lys79)
Unconjugated
Affinity Purified
Rabbit
Affinity Purfied
2-8°C for 12 months
Polyclonal
Human
Immunoassay, Multiplexing, Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
HIST3H3; H3FT; H3t; H3T; H3/g; H3.4; H3/t
Linear Peptide corresponding to human Histone H3 methylated at Lys79.
100μg
Primary
NP_003484.1