Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trimethyl Histone H3(Lys9) Mouse anti-Human, Vertebrates, Clone: CMA308, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051250
Description
Anti-Trimethyl Histone H3(Lys9) Antibody, clone CMA308 is a mouse monoclonal antibody for detection of Trimethyl Histone H3(Lys9) also known as H3K9me3, Histone H3 (tri methyl K9) and has been validated in western blot, ELISA, immunocytochemistry, immunoprecipitation, Mplex.Specifications
Trimethyl Histone H3(Lys9) | |
Monoclonal | |
Unconjugated | |
NM_003493 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 κ |
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
CMA308 | |
Purified mouse monoclonal IgG1κ in PBS with 0.05% sodium azide. | |
H3., H3/g, H3/t , H3FT, H3T , H3t , MGC126886, MGC126888, OTTHUMP00000037945 | |
Synthetic peptide corresponding to amino acids 1-19 of human Histone H3, trimethylated on Lys9, conjugated to KLH. | |
100 μg | |
Human, Vertebrates | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only