Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TrkA Sheep anti-Rat, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Manufacturer:  Invitrogen OST00116G

Catalog No. OST00116G


Add to cart

Description

Description

Glycerol (1:1) may be added for additional stability. Reconstitute in 500 µL of sterile water.

TrkA (tropomyosin receptor kinase A) encodes a member of the neurotrophic tyrosine kinase receptor (NTKR) family. TrkA is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. The presence of TrkA leads to cell differentiation and may play a role in specifying sensory neuron subtypes. Mutations in TrkA have been associated with congenital insensitivity to pain, anhidrosis, self-mutilating behavior, mental retardation and cancer. Alternate transcriptional splice variants of this gene have been found, but only three have been characterized to date. Mutations in the TRKA gene have been associated with congenital insensitivity to pain, anhidrosis, self-mutilating behavior, mental retardation and cancer.
Specifications

Specifications

TrkA
Polyclonal
Unconjugated
Ntrk1
Lyophilized
NTRK1
A synthetic peptide from the extracellular domain of rat TrkA.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Rat
Immunohistochemistry, Western Blot
1 mg/mL
Antibody with no preservative
P35739
High affinity nerve growth factor receptor, NTRK1, p140-TrkA, Trk-A, Neurotrophic tyrosine kinase receptor type 1, Slow nerve growth factor receptor
Sheep
IgG
500 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
59109
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit