Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trop-2 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABC425
Description
Anti-Trop-2 Antibody detects level of Trop-2 and has been published and validated for use in Trop-2.
Specifications
Trop-2 | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Protein G purified | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Western Blot | |
Unconjugated | |
TACSTD2; GA733-1; M1S1; TROP2 | |
Linear peptide corresponding to the intracellular domain of human Trop-2. | |
100 μg | |
NP_002344 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction