Learn More
anti-Troponin T type 1 (slow skeletal), Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP132748
Catalog No. NBP132748
Add to cart
Description
Description
Troponin T type 1 (slow skeletal) Polyclonal antibody specifically detects Troponin T type 1 (slow skeletal) in Human, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Troponin T type 1 (slow skeletal)
0.89mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
TNNT1
Full length human Troponin T1 (slow skeletal) Recombinant protein.
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
ANMtroponin T, slow skeletal muscle, FLJ98147, MGC104241, Slow skeletal muscle troponin T, STNT, TNTS, TNTtroponin T1, skeletal, slow, troponin T type 1 (skeletal, slow), troponin-T1, skeletal, slow
Rabbit
IgG
0.1 ml
Cytoskeleton Markers, Stem Cell Markers
Primary
7138
Human, Porcine